missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MICA/B Monoclonal Antibody (6D4), Biotin

Product Code. 17491033
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17491033 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17491033 Supplier Invitrogen™ Supplier No. MA538726

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

This antibody recognizes a common extracellular epitope on MICA and MICB glycoproteins, transmembrane ligands of CD314 (NKG2D), and is able to block CD314-mediated activation of NK cells and cytotoxic T cells.

MICA encodes the higly polymorphic MHC (HLA) class I chain-related gene A. The protein product is expressed on the cell surface, although unlike canonical class I molecules does not seem to associate with beta-2-microglobulin. It is thought that MICA functions as a stress-induced antigen that is broadly recognized by intestinal epithelial gamma delta T cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MICA/B
Applications Flow Cytometry, Immunoprecipitation, Immunocytochemistry
Classification Monoclonal
Clone 6D4
Concentration 1 mg/mL
Conjugate Biotin
Formulation PBS with 15 mM sodium azide; pH 7.4
Gene MICA
Gene Accession No. Q29980, Q29983
Gene Alias HLA class I antigen; MHC class I chain-related protein A; MHC class I chain-related protein B; MHC class I mic-B antigen; MHC class I polypeptide-related sequence A; MHC class I polypeptide-related sequence B; MHC class I related sequence A; MHC class I-like molecule PERB11.2-IMX; MICA; MIC-A; MICB; MIC-B; PERB11.1; PERB11.2; sMICA; sMICB; soluble MICA; soluble MICB; stress inducible class I homolog; truncated MHC class I polypeptide-related sequence A
Gene Symbols MICA, MICB
Host Species Mouse
Immunogen Transfected C1R cells expressing MICA.
Purification Method Size-exclusion chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100507436, 4277
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.