Learn More
Merck Millicell HY 3-layer Cell Culture Flask
Shop All Merck Products
Click to view available options
:
Culture area: 600cm2
Description
- Consistent, high cell yields and viability
- Same media requirements (volume per unit area) as traditional T-flasks
- Uniform culture conditions without media leaking or spillover between layers
Specifications
Specifications
| Material | Polystyrene |
| Height (Metric) | 32 mm |
| Length (Metric) | 222 mm |
| No. of Positions | 3 layers |
| No. per Case | 16 |
| No. per Pack | 2/Bag |
| Temperature Range (Metric) | 45°C Max. |
| Width (Metric) | 12 cm |
| Sterility | Sterile |
Product Content Correction
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto