Learn More
Milrinone, 98+%, Thermo Scientific™
Potent cAMP-specific phosphodiesterase inhibitor
144.00€ - 496.00€
Chemical Identifiers
| CAS | 78415-72-2 |
|---|---|
| Molecular Formula | C12H9N3O |
| Molecular Weight (g/mol) | 211.224 |
| MDL Number | MFCD00133539 |
| InChI Key | PZRHRDRVRGEVNW-UHFFFAOYSA-N |
| Synonym | milrinone, primacor, corotrope, corotrop, milrinona, milrinonum, milrinonum latin, milrinona spanish, milrila, unii-ju9yax04c7 |
| PubChem CID | 4197 |
| ChEBI | CHEBI:50693 |
| IUPAC Name | 6-methyl-2-oxo-5-pyridin-4-yl-1H-pyridine-3-carbonitrile |
| SMILES | CC1=C(C=C(C(=O)N1)C#N)C2=CC=NC=C2 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15424989
|
Thermo Scientific Alfa Aesar
J62659.MA |
10mg |
151.00€ 144.00€ / 10mg Save 7.00€ 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15444989
|
Thermo Scientific Alfa Aesar
J62659.MF |
50mg |
506.00€ 496.00€ / 50mg Save 10.00€ 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is used (particularly intravenously, as the lactate) for the short-term management of severe heart failure. Milrinone inhibits the action of phosphodiesterase-3 preventing degradation of cAMP. The corresponding increase in cAMP levels leads to increased activation of protein kinase A. It is a selective phosphodiesterase inhibitor with vasodilating and positive inotropic activity.
Solubility
Soluble in DMSO. Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 78415-72-2 | |
| 211.224 | |
| PZRHRDRVRGEVNW-UHFFFAOYSA-N | |
| 4197 | |
| 6-methyl-2-oxo-5-pyridin-4-yl-1H-pyridine-3-carbonitrile |
| C12H9N3O | |
| MFCD00133539 | |
| milrinone, primacor, corotrope, corotrop, milrinona, milrinonum, milrinonum latin, milrinona spanish, milrila, unii-ju9yax04c7 | |
| CHEBI:50693 | |
| CC1=C(C=C(C(=O)N1)C#N)C2=CC=NC=C2 |
Specifications
| 78415-72-2 | |
| MFCD00133539 | |
| 14,6197 | |
| Soluble in DMSO; Insoluble in water. | |
| CC1=C(C=C(C(=O)N1)C#N)C2=CC=NC=C2 | |
| 211.224 | |
| CHEBI:50693 | |
| ≥98% |
| C12H9N3O | |
| UN2811 | |
| milrinone, primacor, corotrope, corotrop, milrinona, milrinonum, milrinonum latin, milrinona spanish, milrila, unii-ju9yax04c7 | |
| PZRHRDRVRGEVNW-UHFFFAOYSA-N | |
| 6-methyl-2-oxo-5-pyridin-4-yl-1H-pyridine-3-carbonitrile | |
| 4197 | |
| 211.22 | |
| Milrinone |
Safety and Handling
GHS H Statement
H300-H311-H331
Fatal if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
H301+H311+H331
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 278-903-6
missing translation for 'rtecsNumber' : DW1762000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.