missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MIPOL1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15749720 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15749720 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15749720 Supplier Invitrogen™ Supplier No. PA551765

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: KTAALVEEVY FAQKERDEAV MSRLQLAIEE RDEAIARAKH MEMSLKVLEN INPEENDMTL QELLNRINNA DTGIAIQKNG AIIVDRIYKT KECKMRITAE EMSALIEERD AALSKCKRLE QELHHVKEQN QTSANNMRHL TAENNQ Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 88%.

This gene encodes a coiled-coil domain-containing protein. The encoded protein may function as a tumor suppressor. A translocation that results in truncation of the protein encoded by this locus has been associated with mirror-image polydactyly, also known as Laurin-Sandrow Syndrome. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIPOL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MIPOL1
Gene Accession No. Q8TD10
Gene Alias 1700081O04Rik; 6030439O22Rik; AU021838; AW536152; CCDC193; CDC91L1; D12Ertd19e; MIPOL1; mirror-image polydactyly 1; mirror-image polydactyly gene 1 homolog (human); mirror-image polydactyly gene 1 protein; mirror-image polydactyly gene 1 protein homolog
Gene Symbols MIPOL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MIPOL1. Recombinant protein control fragment (Product #RP-89055).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145282
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.