missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-17 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94898-0.02ml
This item is not returnable.
View return policy
Description
MMP-17 Polyclonal antibody specifically detects MMP-17 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| MMP-17 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| matrix metallopeptidase 17 (membrane-inserted), matrix metalloproteinase 17 (membrane-inserted), matrix metalloproteinase-17, membrane-type matrix metalloproteinase 4, membrane-type-4 matrix metalloproteinase, MMP-17, MT4MMP, MT4-MMP, MT-MMP 4, MTMMP4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 265-565 of human MMP-17 (NP_057239.4). IMRPYYQGPVGDPLRYGLPYEDKVRVWQLYGVRESVSPTAQPEEPPLLPEPPDNRSSAPPRKDVPHRCSTHFDAVAQIRGEAFFFKGKYFWRLTRDRHLVSLQPAQMHRFWRGLPLHLDSVDAVYERTSDHKIVFFKGDRYWVFKDNNVEEGYPRPVSDFSLPPGGIDAAFSWAHNDRTYFFKDQLYWRYDDHTRHMDPGYPAQSPLWRGVPSTLDDAMRWSDGASYFFRGQEYWKVLDGELEVAPGYPQSTARDWLVCGDSQADGSVAAGVDAAEGPRAPPGQHDQSRSEDGYEVCSCTS | |
| 0.02 mL | |
| Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 4326 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction