missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-17 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
232.00€ - 470.00€
Specifications
| Antigen | MMP-17 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18631862
|
Novus Biologicals
NBP2-94898-0.02ml |
0.02 mL |
232.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18696922
|
Novus Biologicals
NBP2-94898-0.1ml |
0.1 mL |
470.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MMP-17 Polyclonal antibody specifically detects MMP-17 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| MMP-17 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 4326 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| matrix metallopeptidase 17 (membrane-inserted), matrix metalloproteinase 17 (membrane-inserted), matrix metalloproteinase-17, membrane-type matrix metalloproteinase 4, membrane-type-4 matrix metalloproteinase, MMP-17, MT4MMP, MT4-MMP, MT-MMP 4, MTMMP4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 265-565 of human MMP-17 (NP_057239.4). IMRPYYQGPVGDPLRYGLPYEDKVRVWQLYGVRESVSPTAQPEEPPLLPEPPDNRSSAPPRKDVPHRCSTHFDAVAQIRGEAFFFKGKYFWRLTRDRHLVSLQPAQMHRFWRGLPLHLDSVDAVYERTSDHKIVFFKGDRYWVFKDNNVEEGYPRPVSDFSLPPGGIDAAFSWAHNDRTYFFKDQLYWRYDDHTRHMDPGYPAQSPLWRGVPSTLDDAMRWSDGASYFFRGQEYWKVLDGELEVAPGYPQSTARDWLVCGDSQADGSVAAGVDAAEGPRAPPGQHDQSRSEDGYEVCSCTS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title