Learn More
MMR/CD206/Mannose Receptor Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 8 publications
Brand:  Novus Biologicals NBP1-90020
 View more versions of this product
Product Code. 18039625
  • 603.00€ / 0.10mL
Description
MMR/CD206/Mannose Receptor Polyclonal specifically detects MMR/CD206/Mannose Receptor in Human, Mouse, Porcine, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MMR/CD206/Mannose Receptor
Polyclonal
Western Blot 0.04-0.4 ug/ml, Flow Cytometry Validated for Flow from a verified customer review., Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence Reported in scientific literature (PMID 28620274)., Immunohistochemistry-Paraffin 1:1000 - 1:2500
CD206, CLEC13Dmacrophage mannose receptor 1, C-type lectin domain family 13 member D, mannose receptor, C type 1, MMRCD206 antigen
Rabbit
Affinity Purified
RUO
4360
Human, Mouse, Pig
IgG
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
MRC1
This antibody was developed against Recombinant Protein corresponding to amino acids:NEDHKRCVDAVSPSAVQTAACNQDAESQKFRWVSESQIMSVAFKLCLGVPSKTDWVAITLYACDSKSEFQKWECKNDTLLGIKGEDLFFNYGNRQEKNIMLYKGSGLWSRWKIYG
0.1 mL
Primary
Specificity of human MMR/CD206/Mannose Receptor antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.