missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMR/CD206/Mannose Receptor Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 8 publications
265.00€ - 603.00€
Specifications
| Antigen | MMR/CD206/Mannose Receptor |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/ml, Flow Cytometry Validated for Flow from a verified customer review., Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence Reported in scientific literature (PMID 28620274)., Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18465212
|
Novus Biologicals
NBP1-90020-25ul |
25 μL |
265.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18039625
|
Novus Biologicals
NBP1-90020 |
0.1 mL |
603.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MMR/CD206/Mannose Receptor Polyclonal specifically detects MMR/CD206/Mannose Receptor in Human, Mouse, Porcine, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| MMR/CD206/Mannose Receptor | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 4360 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:NEDHKRCVDAVSPSAVQTAACNQDAESQKFRWVSESQIMSVAFKLCLGVPSKTDWVAITLYACDSKSEFQKWECKNDTLLGIKGEDLFFNYGNRQEKNIMLYKGSGLWSRWKIYG | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.04-0.4 ug/ml, Flow Cytometry Validated for Flow from a verified customer review., Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence Reported in scientific literature (PMID 28620274)., Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Pig | |
| CD206, CLEC13Dmacrophage mannose receptor 1, C-type lectin domain family 13 member D, mannose receptor, C type 1, MMRCD206 antigen | |
| MRC1 | |
| IgG | |
| Affinity Purified | |
| Specificity of human MMR/CD206/Mannose Receptor antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title