missing translation for 'onlineSavingsMsg'
Learn More

mNeonGreen Mouse, Unconjugated, Clone: 32F6, Proteintech ChromoTek

Product Code. p-7235969
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17353263 20 μL 20µL
17343263 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17353263 Supplier Chromotek Supplier No. 32f620

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mouse monoclonal antibody [32F6] to mNeonGreen fluorescent protein derived from Branchiostoma lanceolatum.

Specifications

Antigen mNeonGreen
Applications Western Blot, Immunofluorescence, ELISA
Classification Monoclonal
Clone 32F6
Conjugate Unconjugated
Host Species Mouse
Quantity 20 μL
Primary or Secondary Primary
Content And Storage Shipped at ambient temperature. Store at +4°C. Stable for one year.
Isotype IgG2c
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.