missing translation for 'onlineSavingsMsg'
Learn More

Mono(4-hydroxybutyl) Terephthalate (>90%), TRC

Product Code. 30138555
Change view
Click to view available options
Quantity:
10 mg
250 mg
Unit Size:
10mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138555 250 mg 250mg
30008952 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138555 Supplier Toronto Research Chemicals Supplier No. TRCM547530250MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63317-89-5
Molecular Formula C12 H14 O5
Molecular Weight (g/mol) 238.24
Synonym 1-(4-Hydroxybutyl) Ester 1,4-benzenedicarboxylic Acid
IUPAC Name 4-(4-hydroxybutoxycarbonyl)benzoic acid
SMILES OCCCCOC(=O)c1ccc(cc1)C(=O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H14O5/c13-7-1-2-8-17-12(16)10-5-3-9(4-6-10)11(14)15/h3-6,13H,1-2,7-8H2,(H,14,15)
Formula Weight 238.0841
Chemical Name or Material Mono(4-hydroxybutyl) Terephthalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.