missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Monomethyl auristatin F (MMAF) Monoclonal antibody specifically detects Monomethyl auristatin F (MMAF) in Nonspecies specific samples. It is validated for ELISA
Specifications
Specifications
| Antigen | Monomethyl auristatin F (MMAF) |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 01 |
| Conjugate | Unconjugated |
| Dilution | ELISA 1:1000-1:2000 |
| Formulation | 0.2 um filtered solution in PBS |
| Gene Alias | 745017-94-1, mafodotin, MMAF (GMP) |
| Host Species | Mouse |
| Immunogen | This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with Monomethyl auristatin F (MMAF) conjugated to BSA |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?