missing translation for 'onlineSavingsMsg'
Learn More

D-dimer (Fibrin Degradation Product), Mouse, Biotin, Clone: DD41, Abnova™

Product Code. 16036846
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16036846 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16036846 Supplier Abnova Supplier No. MAB5487.1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against D-dimer (Fibrin Degradation Product).

Specifications

Antigen D-dimer (Fibrin Degradation Product)
Applications Western Blot
Classification Monoclonal
Clone B2654M
Conjugate Biotin
Description Mouse monoclonal antibody raised against D-dimer (Fibrin Degradation Product).
Dilution The optimal working dilution should be determined by the end user.
Formulation In 20mM Tris-HCl, 150mM NaCl, pH 7.5 (0.09% sodium azide)
Host Species Mouse
Immunogen Mixture of D-dimer and high molecular weight fibrin degradation products.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes high molecular weight fibrin degradation products. Recognizes D-dimer in Western Blot under non-reducing conditions. Interacts with beta-chain of D-dimer in Western Blot under reducing conditions. Does not cross-react with fibrinogen and D-monomer.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.