missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Artemin Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB10851-100
This item is not returnable.
View return policy
Description
Artemin Monoclonal specifically detects Artemin in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Artemin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 3-25 ug/mL | |
| Q9Z0L2.1 | |
| ARTN | |
| E. coli-derived recombinant mouse Artemin Ala112-Gly224 Accession # Q9Z0L2.1 | |
| 100 μg | |
| Primary | |
| Detects mouse Artemin in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) AgRP, rmCripto, recombinant human (rh) Cripto-1, recombinant drosophila Dpp, recombinant rat GDNF, rhLAP, rhLatent TGF-beta 1, rhLefty A, rmLefty-1, rmMIS, rhNeurturin, rmNodal, rhTGF-alpha , rhTGF-beta 1, rhTGF-beta 1.2, rhTGF-beta 2, rhTGF-beta 3, or recombinant Amphibian TGF-beta 5 is observed. | |
| Mouse | |
| Supernatant |
| Western Blot, Immunohistochemistry | |
| 185234 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| artemin, ARTN, ENOVIN, EVN, EVNneurotrophic factor, NBN, Neublastin | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9048 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction