missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Artemin Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
201.00€ - 1675.00€
Specifications
| Antigen | Artemin |
|---|---|
| Clone | 185234 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 3-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18752477
![]() |
R&D Systems
MAB10851-SP |
25 μg |
201.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18784438
![]() |
R&D Systems
MAB10851-100 |
100 μg |
416.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18702947
![]() |
R&D Systems
MAB10851-500 |
500 μg |
1675.00€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
Artemin Monoclonal specifically detects Artemin in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Especificaciones
| Artemin | |
| LYOPH | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| artemin, ARTN, ENOVIN, EVN, EVNneurotrophic factor, NBN, Neublastin | |
| ARTN | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 185234 | |
| Western Blot 1 ug/mL, Immunohistochemistry 3-25 ug/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| Q9Z0L2.1 | |
| 9048 | |
| E. coli-derived recombinant mouse Artemin Ala112-Gly224 Accession # Q9Z0L2.1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Artemin in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) AgRP, rmCripto, recombinant human (rh) Cripto-1, recombinant drosophila Dpp, recombinant rat GDNF, rhLAP, rhLatent TGF-beta 1, rhLefty A, rmLefty-1, rmMIS, rhNeurturin, rmNodal, rhTGF-alpha , rhTGF-beta 1, rhTGF-beta 1.2, rhTGF-beta 2, rhTGF-beta 3, or recombinant Amphibian TGF-beta 5 is observed. |
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto