missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Cathepsin D Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 9 publications
Brand: R&D Systems MAB1029-SP
This item is not returnable.
View return policy
Description
Cathepsin D Monoclonal specifically detects Cathepsin D in Mouse samples. It is validated for Western Blot.
Specifications
| Cathepsin D | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P18242 | |
| CTSD | |
| Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin D Ile21-Leu410 Accession # P18242 | |
| 25 μg | |
| Primary | |
| Detects both the pro and active forms of mouse Cathepsin D in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Cathepsin D or recombinant mouse Cathepsin E is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 204712 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cathepsin D, cathepsin D (lysosomal aspartyl protease), CPSD, CTSD, EC 3.4.23, EC 3.4.23.5, lysosomal aspartyl peptidase, lysosomal aspartyl protease, MGC2311, neuronal 10 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1509 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction