missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Cathepsin D Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 9 publications
214.00€ - 584.00€
Specifications
| Antigen | Cathepsin D |
|---|---|
| Clone | 204712 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18714448
![]() |
R&D Systems
MAB1029-SP |
25 μg |
214.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18130271
![]() |
R&D Systems
MAB1029 |
500 μg |
584.00€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cathepsin D Monoclonal specifically detects Cathepsin D in Mouse samples. It is validated for Western Blot.Specifications
| Cathepsin D | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| P18242 | |
| 1509 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin D Ile21-Leu410 Accession # P18242 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects both the pro and active forms of mouse Cathepsin D in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Cathepsin D or recombinant mouse Cathepsin E is observed. |
| 204712 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cathepsin D, cathepsin D (lysosomal aspartyl protease), CPSD, CTSD, EC 3.4.23, EC 3.4.23.5, lysosomal aspartyl peptidase, lysosomal aspartyl protease, MGC2311, neuronal 10 | |
| CTSD | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title