missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse EphA4 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB641
This item is not returnable.
View return policy
Description
EphA4 Monoclonal specifically detects EphA4 in Mouse samples. It is validated for Western Blot.
Specifications
| EphA4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cek8, EC 2.7.10, EC 2.7.10.1, EK8, EPH receptor A4, EphA4, EPH-like kinase 8, ephrin type-A receptor 4, HEK8, receptor protein-tyrosine kinase HEK8, SEK, Sek1, Tyro1, TYRO1 protein tyrosine kinase, Tyrosine-protein kinase receptor SEK, Tyrosine-protein kinase TYRO1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2043 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 107925 | |
| Western Blot 1 ug/mL | |
| Q03137 | |
| EPHA4 | |
| Mouse myeloma cell line NS0-derived recombinant mouse EphA4 Val20-Thr547 Accession # Q03137 | |
| 500 μg | |
| Primary | |
| Detects mouse EphA4 in direct ELISAs and Western blots. Shows less than 5% cross-reactivity with recombinant mouse (rm) EphA3 and recombinant rat (rr) EphB1 and no cross-reactivity with recombinant human EphA1, rmEphA2, -A6, -A7, -A8, -B2, -B3, -B4, -B6, or rrEphA5. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction