missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse EphA4 Antibody, R&D Systems™

Rat Monoclonal Antibody
209.00€ - 532.00€
Specifications
| Antigen | EphA4 |
|---|---|
| Clone | 107925 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18760728
![]() |
R&D Systems
MAB641-SP |
25 μg |
209.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18122881
|
R&D Systems
MAB641 |
500 μg |
532.00€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EphA4 Monoclonal specifically detects EphA4 in Mouse samples. It is validated for Western Blot.Specifications
| EphA4 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cek8, EC 2.7.10, EC 2.7.10.1, EK8, EPH receptor A4, EphA4, EPH-like kinase 8, ephrin type-A receptor 4, HEK8, receptor protein-tyrosine kinase HEK8, SEK, Sek1, Tyro1, TYRO1 protein tyrosine kinase, Tyrosine-protein kinase receptor SEK, Tyrosine-protein kinase TYRO1 | |
| EPHA4 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 107925 | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q03137 | |
| 2043 | |
| Mouse myeloma cell line NS0-derived recombinant mouse EphA4 Val20-Thr547 Accession # Q03137 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse EphA4 in direct ELISAs and Western blots. Shows less than 5% cross-reactivity with recombinant mouse (rm) EphA3 and recombinant rat (rr) EphB1 and no cross-reactivity with recombinant human EphA1, rmEphA2, -A6, -A7, -A8, -B2, -B3, -B4, -B6, or rrEphA5. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title