missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse ESAM Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB28271
This item is not returnable.
View return policy
Description
ESAM Monoclonal specifically detects ESAM in Mouse samples. It is validated for Immunohistochemistry.
Specifications
| ESAM | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| Q925F2 | |
| ESAM | |
| Mouse myeloma cell line NS0-derived recombinant mouse ESAM Gln30-Ser248 (predicted) Accession # Q925F2 | |
| 100 μg | |
| Primary | |
| Detects mouse ESAM in direct ELISAs. | |
| Mouse | |
| Purified |
| Immunohistochemistry | |
| 340236 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 2310008D05Rik, endothelial cell adhesion molecule, endothelial cell-selective adhesion molecule, HUEL (C4orf1)-interacting protein, LP4791 protein, W117m | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 90952 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction