missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse ESAM Antibody, R&D Systems™

Rat Monoclonal Antibody
209.00€ - 396.00€
Specifications
| Antigen | ESAM |
|---|---|
| Clone | 340236 |
| Concentration | LYOPH |
| Dilution | Immunohistochemistry 8-25 ug/mL |
| Applications | Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18719427
![]() |
R&D Systems
MAB28271-SP |
25 μg |
209.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18129380
![]() |
R&D Systems
MAB28271 |
100 μg |
396.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ESAM Monoclonal specifically detects ESAM in Mouse samples. It is validated for Immunohistochemistry.Specifications
| ESAM | |
| LYOPH | |
| Immunohistochemistry | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q925F2 | |
| 90952 | |
| Mouse myeloma cell line NS0-derived recombinant mouse ESAM Gln30-Ser248 (predicted) Accession # Q925F2 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse ESAM in direct ELISAs. |
| 340236 | |
| Immunohistochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 2310008D05Rik, endothelial cell adhesion molecule, endothelial cell-selective adhesion molecule, HUEL (C4orf1)-interacting protein, LP4791 protein, W117m | |
| ESAM | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel