missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse HPRG Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1905-SP
This item is not returnable.
View return policy
Description
HPRG Polyclonal specifically detects HPRG in Mouse samples. It is validated for Western Blot.
Specifications
| HPRG | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp779H1622, Histidine-proline-rich glycoprotein, histidine-rich glycoprotein, HPRGHRGP, HRG | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3273 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| BAB33094 | |
| HRG | |
| Mouse myeloma cell line NS0-derived recombinant mouse HPRG Leu19-Lys525 Accession # BAB33094 | |
| 25 μg | |
| Primary | |
| Detects mouse HPRG in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human HPRG is observed and less than 2% cross-reactivity wth recombinant mouse (rm) Cystatin A, rmCystatin B, rmCystatin C, and rmCystatin E/M is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction