missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse HPRG Antibody, R&D Systems™

Goat Polyclonal Antibody
209.00€ - 480.00€
Specifications
| Antigen | HPRG |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18789117
![]() |
R&D Systems
AF1905-SP |
25 μg |
209.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18011402
|
R&D Systems
AF1905 |
100 μg |
480.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HPRG Polyclonal specifically detects HPRG in Mouse samples. It is validated for Western Blot.Specifications
| HPRG | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| DKFZp779H1622, Histidine-proline-rich glycoprotein, histidine-rich glycoprotein, HPRGHRGP, HRG | |
| HRG | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| BAB33094 | |
| 3273 | |
| Mouse myeloma cell line NS0-derived recombinant mouse HPRG Leu19-Lys525 Accession # BAB33094 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse HPRG in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human HPRG is observed and less than 2% cross-reactivity wth recombinant mouse (rm) Cystatin A, rmCystatin B, rmCystatin C, and rmCystatin E/M is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title