missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse PCPE-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2239-SP
This item is not returnable.
View return policy
Description
PCPE-1/PCOLCE Monoclonal specifically detects PCPE-1/PCOLCE in Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| PCPE-1/PCOLCE | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PCOLCE, PCPE, PCPE1, PCPE-1, PCPE1Type I procollagen COOH-terminal proteinase enhancer, procollagen C-endopeptidase enhancer, procollagen C-endopeptidase enhancer 1, Procollagen COOH-terminal proteinase enhancer 1, Procollagen C-proteinase enhancer 1, procollagen, type 1, COOH-terminal proteinase enhancer, Type 1 procollagen C-proteinase enhancer protein | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5118 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunoprecipitation | |
| 261845 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q61398 | |
| PCOLCE | |
| Mouse myeloma cell line NS0-derived recombinant mouse PCPE-1 Gln25-Ala468 Accession # Q61398 | |
| 25 μg | |
| Primary | |
| Detects mouse PCPE-1 in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant human PCPE-1 is observed. In Western blots, no cross-reactivity with recombinant human PCPE-1 is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction