missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse PCPE-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
201.00€ - 537.00€
Specifications
| Antigen | PCPE-1/PCOLCE |
|---|---|
| Clone | 261845 |
| Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18784469
![]() |
R&D Systems
MAB2239-SP |
25 μg |
201.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18193001
![]() |
R&D Systems
MAB2239 |
500 μg |
537.00€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PCPE-1/PCOLCE Monoclonal specifically detects PCPE-1/PCOLCE in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| PCPE-1/PCOLCE | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PCOLCE, PCPE, PCPE1, PCPE-1, PCPE1Type I procollagen COOH-terminal proteinase enhancer, procollagen C-endopeptidase enhancer, procollagen C-endopeptidase enhancer 1, Procollagen COOH-terminal proteinase enhancer 1, Procollagen C-proteinase enhancer 1, procollagen, type 1, COOH-terminal proteinase enhancer, Type 1 procollagen C-proteinase enhancer protein | |
| PCOLCE | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 261845 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q61398 | |
| 5118 | |
| Mouse myeloma cell line NS0-derived recombinant mouse PCPE-1 Gln25-Ala468 Accession # Q61398 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse PCPE-1 in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant human PCPE-1 is observed. In Western blots, no cross-reactivity with recombinant human PCPE-1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title