missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TSLP Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB5551-SP
This item is not returnable.
View return policy
Description
TSLP Monoclonal specifically detects TSLP in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| TSLP | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| thymic stromal lymphopoietin | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 85480 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, ELISA | |
| 152640 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q9JIE6 | |
| TSLP | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse TSLP Tyr20-Glu140 Accession # Q9JIE6 | |
| 25 μg | |
| Primary | |
| Detects mouse TSLP in ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse IL-7 or recombinant human TSLP. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction