missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TSLP Antibody, R&D Systems™

Rat Monoclonal Antibody
201.00€ - 2050.00€
Especificaciones
| Antigen | TSLP |
|---|---|
| Clone | 152640 |
| Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
18765099
![]() |
R&D Systems
MAB5551-SP |
25 μg |
201.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18770939
![]() |
R&D Systems
MAB5551-100 |
100 μg |
512.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18793819
![]() |
R&D Systems
MAB5551-500 |
500 μg |
2050.00€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
TSLP Monoclonal specifically detects TSLP in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Especificaciones
| TSLP | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| Q9JIE6 | |
| 85480 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse TSLP Tyr20-Glu140 Accession # Q9JIE6 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse TSLP in ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse IL-7 or recombinant human TSLP. |
| 152640 | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| thymic stromal lymphopoietin | |
| TSLP | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto