Learn More
MPPE1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16824444 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16824444 150 μL 150µL
16834444 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16824444 Supplier Proteintech Supplier No. 153131AP150UL
Rabbit Polyclonal Antibody
Metallophosphoesterase required for transport of GPI- anchor proteins from the endoplasmic reticulum to the Golgi. Acts in lipid remodeling steps of GPI-anchor maturation by mediating the removal of a side-chain ethanolamine-phosphate (EtNP) from the second Man (Man2) of the GPI intermediate, an essential step for efficient transport of GPI-anchor proteins.
Specifications
Antigen MPPE1
Applications Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene MPPE1
Gene Accession No. B1WC86, Q53F39, Q80XL7
Gene Alias metallophosphoesterase 1, MPPE1, PGAP5
Gene Symbols Mppe1
Host Species Rabbit
Immunogen MPPE1 Fusion Protein Ag7547
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225651, 361344, 65258
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG