Description
Metallophosphoesterase required for transport of GPI- anchor proteins from the endoplasmic reticulum to the Golgi. Acts in lipid remodeling steps of GPI-anchor maturation by mediating the removal of a side-chain ethanolamine-phosphate (EtNP) from the second Man (Man2) of the GPI intermediate, an essential step for efficient transport of GPI-anchor proteins.
Specifications
Specifications
| Antigen | MPPE1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.33 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | MPPE1 |
| Gene Accession No. | B1WC86, Q53F39, Q80XL7 |
| Gene Alias | metallophosphoesterase 1, MPPE1, PGAP5 |
| Gene Symbols | Mppe1 |
| Show More |