Learn More
MRPL19 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16517-1-AP-20UL
 View more versions of this product
Product Code. 16816174
  • 145.00€ / 20µL
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein.
Specifications
Specifications
MRPL19
Polyclonal
Unconjugated
MRPL19
KIAA0104, L15mt, L19mt, MRP L15, MRP L19, MRPL15, MRPL19, RLX1, RPML15
Rabbit
Antigen Affinity Chromatography
RUO
297372, 56284, 9801
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P49406, Q9D338
Mrpl19
MRPL19 Fusion Protein Ag9738
20 μL
Primary
Mouse, Human, Rat