missing translation for 'onlineSavingsMsg'
Learn More

MRPL19 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Specifications

Antigen MRPL19
Concentration 0.15 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16816174
View Documents
Proteintech
16517-1-AP-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16806174
View Documents
Proteintech
16517-1-AP-150UL
150 μL
410.00€
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein.
Specifications

Specifications

MRPL19
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence
Unconjugated
Rabbit
Mouse, Human, Rat
P49406, Q9D338
297372, 56284, 9801
MRPL19 Fusion Protein Ag9738
Primary
-20°C
MRPL19
0.15 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
KIAA0104, L15mt, L19mt, MRP L15, MRP L19, MRPL15, MRPL19, RLX1, RPML15
Mrpl19
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.