missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MRPL3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15969014
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15969014 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15969014 Supplier Invitrogen™ Supplier No. PA566306

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PGAVATGDIGR VWPGTKMPGK MGNIYRTEYG LKVWRINTKH NIIYVNGSVP GHKNCLVKVK DSKLPAYKDL GKNLPFPTYF PDGDEEELPE DLYDENVCQP GA Highest antigen sequence identity to the following orthologs - mouse 83%, rat 83%.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein that belongs to the L3P ribosomal protein family. A pseudogene corresponding to this gene is found on chromosome 13q.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MRPL3
Gene Accession No. P09001
Gene Alias 2010320L16Rik; 39S ribosomal protein L3, mitochondrial; 5930422H18Rik; AA407889; COXPD9; L3mt; mitochondrial 60S ribosomal protein L3; Mitochondrial large ribosomal subunit protein uL3m; mitochondrial ribosomal protein L3; MRL3; Mrpl3; MRP-L3; RPML3
Gene Symbols MRPL3
Host Species Rabbit
Immunogen Recombinant Human MRPL3. Recombinant protein control fragment (Product #RP-106984).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11222
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.