Learn More
MRRF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12357-2-AP-20UL
This item is not returnable.
View return policy
Description
Responsible for the release of ribosomes from messenger RNA at the termination of protein biosynthesis. May increase the efficiency of translation by recycling ribosomes from one round of translation to another By similarity. Ota T., Nat. Genet. 36:40-45(2004). Humphray S.J., Nature 429:369-374(2004). Zhang Y., Biochim. Biophys. Acta 1443:245-250(1998).Specifications
| MRRF | |
| Polyclonal | |
| Unconjugated | |
| MRRF | |
| MRFF, MRRF, MTRRF, RRF | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 311903, 67871, 92399 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5RKI9, Q96E11, Q9D6S7 | |
| Mrrf | |
| MRRF Fusion Protein Ag3021 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |