missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRTO4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20194-1-AP-20UL
This item is not returnable.
View return policy
Description
MRTO4 is a protein sharing a low level of sequence similarity with ribosomal protein P0. While the precise function of the protein is currently unknown, it appears to be involved in mRNA turnover and ribosome assembly.Specifications
| MRTO4 | |
| Polyclonal | |
| Unconjugated | |
| MRTO4 | |
| C1orf33, MRT4, MRTO4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 298586, 51154, 69902 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9D0I8, Q9UKD2 | |
| Mrto4 | |
| MRTO4 Fusion Protein Ag14122 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction