missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRTO4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | MRTO4 |
|---|---|
| Concentration | 0.33 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16819714
|
Proteintech
20194-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16809714
|
Proteintech
20194-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MRTO4 is a protein sharing a low level of sequence similarity with ribosomal protein P0. While the precise function of the protein is currently unknown, it appears to be involved in mRNA turnover and ribosome assembly.Specifications
| MRTO4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| Q9D0I8, Q9UKD2 | |
| 298586, 51154, 69902 | |
| MRTO4 Fusion Protein Ag14122 | |
| Primary | |
| -20°C | |
| MRTO4 |
| 0.33 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| C1orf33, MRT4, MRTO4 | |
| Mrto4 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title