missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MSH2 Monoclonal Antibody (10G1)
GREENER_CHOICE

Product Code. 17825722
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17825722 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17825722 Supplier Invitrogen™ Supplier No. MA545060

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MSH2 Monoclonal Antibody for Western Blot, IHC (P)

Plays a role in neurogenesis and neuronal migration. Necessary for correct formation of mitotic spindles and chromosome separation during mitosis. Necessary for cytokinesis and cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSH2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 10G1
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene Msh2
Gene Accession No. P43246, P54275
Gene Alias AI788990; COCA1; DNA mismatch repair protein MSH2; FCC1; hMSH2; HNPCC; HNPCC1; LCFS2; mismatch repair ATPase MSH2; mismatch repair protein; MSH2; Msh2p; mutS homolog 2; mutS homolog 2 (E. coli); mutS homolog 2, colon cancer, nonpolyposis type 1; mutS protein homolog 2; O0935; PMS5; YOL090W
Gene Symbols Msh2
Host Species Mouse
Immunogen Synthetic peptide within N-terminal human MSH2.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4436, 81709
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.