Learn More
Invitrogen™ MSH4 Polyclonal Antibody

Description
Immunogen sequence: QIMAQIGSYV PAEYSSFRIA KQIFTRISTD DDIETNSSTF MKEMKEIAYI LHNANDKSLI LIDELGRGTN TEEGIGICYA VCEYLLSLKA FTLFATHFLE LCHIDALYPN VENMHFEVQH VKNTSRNKEA ILYTYKLSKG LTEEKNYGLK AAEVSSLPPS IVLDAKEITT QITRQILQNQ RSTPEMERQR AVYHLATRLV QTARNSQLDP DSLRIYLSNL KKKYKEDFPR TEQVPEKTEE; Positive Samples: SKOV3.
Specifications
Specifications
| Antigen | MSH4 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.99 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | MSH4 |
| Gene Accession No. | O15457 |
| Gene Alias | 4930485C04Rik; AV144863; hMSH4; mMsh4; Msh4; mutS homolog 4; mutS homolog 4 (E. coli); MutS homolog 4 variant iota 1; MutS homolog 4 variant theta 1; mutS protein homolog 4 |
| Gene Symbols | MSH4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.