missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Muscarinic Acetylcholine Receptor M2/CHRM2 Antibody (31-1D1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-2805
This item is not returnable.
View return policy
Description
Muscarinic Acetylcholine Receptor M2/CHRM2 Monoclonal specifically detects Muscarinic Acetylcholine Receptor M2/CHRM2 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Muscarinic Acetylcholine Receptor M2/CHRM2 | |
| Monoclonal | |
| 2 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 2 ug/ml, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:20 | |
| CHRM2 | |
| Purified porcine cardiac m2 muscarinic acetylcholine receptor. | |
| 250 μL | |
| GPCR, Neuroscience | |
| 1129 | |
| Human, Mouse, Rat, Pig | |
| Ascites |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 31-1D1 | |
| Unconjugated | |
| 7TM receptor, cholinergic receptor, muscarinic 2, FLJ43243, HM2, MGC120006, MGC120007, muscarinic acetylcholine receptor M2, muscarinic M2 receptor | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| This is specific for the m2 mAChR subtype. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction