missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Muscarinic Acetylcholine Receptor M2/CHRM2 Antibody (31-1D1), Novus Biologicals™
Mouse Monoclonal Antibody
250.00€ - 533.00€
Specifications
| Antigen | Muscarinic Acetylcholine Receptor M2/CHRM2 |
|---|---|
| Clone | 31-1D1 |
| Concentration | 2 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 2 ug/ml, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:20 |
| Applications | Immunofluorescence, Immunocytochemistry, Immunohistochemistry, Flow Cytometry, Immunoprecipitation, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18097174
|
Novus Biologicals
NB120-2805-125UL |
125 μL |
250.00€
125µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18017465
|
Novus Biologicals
NB120-2805 |
250 μL |
533.00€
250µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Muscarinic Acetylcholine Receptor M2/CHRM2 Monoclonal specifically detects Muscarinic Acetylcholine Receptor M2/CHRM2 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| Muscarinic Acetylcholine Receptor M2/CHRM2 | |
| 2 mg/mL | |
| Immunofluorescence, Immunocytochemistry, Immunohistochemistry, Flow Cytometry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| GPCR, Neuroscience | |
| 7TM receptor, cholinergic receptor, muscarinic 2, FLJ43243, HM2, MGC120006, MGC120007, muscarinic acetylcholine receptor M2, muscarinic M2 receptor | |
| CHRM2 | |
| IgG1 | |
| Protein A purified | |
| This is specific for the m2 mAChR subtype. |
| 31-1D1 | |
| Western Blot 1:1000, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 2 ug/ml, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:20 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Mouse, Pig, Human, Rat | |
| 1129 | |
| Purified porcine cardiac m2 muscarinic acetylcholine receptor. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title