missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MyD88 Antibody (4D6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NBP2-27369SS
This item is not returnable.
View return policy
Description
MyD88 Monoclonal specifically detects MyD88 in Human, Mouse, Primate, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular), CyTOF-ready.
Specifications
| MyD88 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 5-7ug/ml, Flow Cytometry 0.5ug/10^6cells∼, Immunocytochemistry/Immunofluorescence 5ug/ml, Flow (Intracellular), CyTOF-ready | |
| MYD88D, myeloid differentiation primary response gene (88), myeloid differentiation primary response protein MyD88 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in Human (isoform 1: 317 amino acids , isoform 3: 264 aa, isoform 4: 204 aa; isoform 5: 159 aa, CRA_a: 304 aa, CRA_b: 309 aa), Gorilla, Chimp, Rhesus Monkey and Orangutan, 82% homologous in Horse, 73% homologous in Dog, 68% homologous in Cat and Mouse (isoform CRA_a: 314 amino acids, CRA_b: 177 amino acids), and 63% homologous in Rat. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF | |
| 4D6 | |
| Unconjugated | |
| Q99836 | |
| MYD88 | |
| A portion of amino acids 50-100 of human MyD88 was used as the immunogen for this antibody. | |
| 0.025 mg | |
| Cytokine Research, Immunology, Innate Immunity, Protein Kinase | |
| 4615 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction