missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MyD88 Antibody (4D6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 5 publications
195.00€ - 588.00€
Specifications
| Antigen | MyD88 |
|---|---|
| Clone | 4D6 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 5-7ug/ml, Flow Cytometry 0.5ug/10^6cells∼, Immunocytochemistry/Immunofluorescence 5ug/ml, Flow (Intracellular), CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18130995
|
Novus Biologicals
NBP2-27369SS |
0.025 mg |
195.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18709123
|
Novus Biologicals
NBP2-27369 |
588.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
MyD88 Monoclonal specifically detects MyD88 in Human, Mouse, Primate, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular), CyTOF-ready.Specifications
| MyD88 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF | |
| Unconjugated | |
| Mouse | |
| Cytokine Research, Immunology, Innate Immunity, Protein Kinase | |
| Q99836 | |
| 4615 | |
| A portion of amino acids 50-100 of human MyD88 was used as the immunogen for this antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 4D6 | |
| Western Blot 5-7ug/ml, Flow Cytometry 0.5ug/10^6cells∼, Immunocytochemistry/Immunofluorescence 5ug/ml, Flow (Intracellular), CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| MYD88D, myeloid differentiation primary response gene (88), myeloid differentiation primary response protein MyD88 | |
| MYD88 | |
| IgG1 κ | |
| Protein G purified | |
| The amino acid sequence used as immunogen is 100% homologous in Human (isoform 1: 317 amino acids , isoform 3: 264 aa, isoform 4: 204 aa; isoform 5: 159 aa, CRA_a: 304 aa, CRA_b: 309 aa), Gorilla, Chimp, Rhesus Monkey and Orangutan, 82% homologous in Horse, 73% homologous in Dog, 68% homologous in Cat and Mouse (isoform CRA_a: 314 amino acids, CRA_b: 177 amino acids), and 63% homologous in Rat. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title