Learn More
MyD88 Antibody (4D6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NBP2-27369
This item is not returnable.
View return policy
Description
MyD88 Monoclonal specifically detects MyD88 in Human, Mouse, Primate, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular), CyTOF-ready.
Specifications
| Western Blot 5-7ug/ml, Flow Cytometry 0.5ug/10^6cells∼, Immunocytochemistry/Immunofluorescence 5ug/ml, Flow (Intracellular), CyTOF-ready | |
| MYD88D, myeloid differentiation primary response gene (88), myeloid differentiation primary response protein MyD88 |