Learn More
MyD88 Antibody (4D6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 5 publications
Brand:  Novus Biologicals NBP2-27369
 View more versions of this product
Product Code. 18709123
  • 588.00€ / 0.10mg
Description
MyD88 Monoclonal specifically detects MyD88 in Human, Mouse, Primate, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular), CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot 5-7ug/ml, Flow Cytometry 0.5ug/10^6cells∼, Immunocytochemistry/Immunofluorescence 5ug/ml, Flow (Intracellular), CyTOF-ready
MYD88D, myeloid differentiation primary response gene (88), myeloid differentiation primary response protein MyD88