missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myelin Protein Zero Antibody (MPZ/7389), Allophycocyanin, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Myelin Protein Zero Monoclonal antibody specifically detects Myelin Protein Zero in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Myelin Protein Zero |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | MPZ/7389 |
| Conjugate | APC |
| Formulation | PBS |
| Gene Alias | Charcot-Marie-Tooth neuropathy 1B, CHM, CMT1, CMT1B, CMT2I, CMT2J, CMT4E, CMTDI3, HMSNIB, MPP, Myelin peripheral protein, myelin protein P0, myelin protein zeroDSS, P0 |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human Myelin Protein Zero protein |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?