missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-D-glucosamine, TRC

Product Code. 30009479
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
Unit Size:
100g
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009479 100 g 100g
30007476 5 g 5g
30004924 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009479 Supplier Toronto Research Chemicals Supplier No. TRCA178000100G
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7512-17-6
Molecular Formula C8 H15 N O6
Molecular Weight (g/mol) 221.208
Synonym D-Glucose, 2-(acetylamino)-2-deoxy-, D-Glucose, 2-acetamido-2-deoxy- (8CI), 2-(Acetylamino)-2-deoxy-D-glucose, 2-Acetamido-2-deoxy-D-glucose, 2-Acetamido-2-deoxyglucose, 2-Acetamido-D-glucose, Acetylglucosamine, Bio-NAG, D-N-Acetylglucosamine, GreenNAG, M
IUPAC Name N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O

Specifications

Quantity 100 g
InChI Formula InChI=1S/C8H15NO6/c1-3(11)9-5-7(13)6(12)4(2-10)15-8(5)14/h4-8,10,12-14H,2H2,1H3,(H,9,11)/t4-,5-,6-,7-,8?/m1/s1
Formula Weight 221.09
Chemical Name or Material N-Acetylglucosamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.