missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylethanolamine, 90+%

Product Code. 11365229
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11365229 100 g 100g
11375229 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11365229 Supplier Thermo Scientific Alfa Aesar Supplier No. A18638.22
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 142-26-7 | C4H9NO2 | 103.121 g/mol

It finds its uses as antistatic agent, foam boosting agent, hair conditioning humectant, skin conditioning surfactant and viscosity controlling agent in cosmetics industry. It is a non-tacky glycerine alternative for use in skin and hair care. It has humectant and lubricant properties for use in shampoos, treatment products and hair conditioners.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its uses as antistatic agent, foam boosting agent, hair conditioning humectant, skin conditioning surfactant and viscosity controlling agent in cosmetics industry. It is a non-tacky glycerine alternative for use in skin and hair care. It has humectant and lubricant properties for use in shampoos, treatment products and hair conditioners.

Solubility
Soluble in water, 1e+006 mg/L @ 25°C (est), soluble in alcohol and ether.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-26-7
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00002836
InChI Key PVCJKHHOXFKFRP-UHFFFAOYSA-N
Synonym n-2-hydroxyethyl acetamide, n-acetylethanolamine, 2-acetamidoethanol, acetamide, n-2-hydroxyethyl, acetylcolamine, 2-acetylaminoethanol, acetamide mea, n-ethanolacetamide, n-acetyl-2-aminoethanol, hydroxyethyl acetamide
PubChem CID 8880
ChEBI CHEBI:74687
IUPAC Name N-(2-hydroxyethyl)acetamide
SMILES CC(=O)NCCO

Specifications

Density 1.12
Boiling Point 151°C to 155°C (5 mmHg)
Flash Point 177°C (351°F)
Linear Formula CH3CONHCH2CH2OH
Refractive Index 1.472
Quantity 100 g
Solubility Information Soluble in water,1e+006mg/L @ 25°C (est),soluble in alcohol and ether.
Formula Weight 103.12
Percent Purity ≥90%
Chemical Name or Material N-Acetylethanolamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.