missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl Di-O-TBS Epinephrine-d3, TRC

Product Code. 30341712
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30341712 25 mg 25mg
30332273 50 mg 50mg
30332174 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30341712 Supplier Toronto Research Chemicals Supplier No. TRCB27642225MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Benzyl Di-O-TBS Epinephrine-d3
Molecular Formula C28 D3 H44 N O3 Si2
Quantity 25 mg
InChI Formula InChI=1S/C28H47NO3Si2/c1-27(2,3)33(8,9)31-25-18-17-23(19-26(25)32-34(10,11)28(4,5)6)24(30)21-29(7)20-22-15-13-12-14-16-22/h12-19,24,30H,20-21H2,1-11H3/i7D3
SMILES [2H]C([2H])([2H])N(CC(O)c1ccc(O[Si](C)(C)C(C)(C)C)c(O[Si](C)(C)C(C)(C)C)c1)Cc2ccccc2
IUPAC Name 2-[benzyl(trideuteriomethyl)amino]-1-[3,4-bis[[tert-butyl(dimethyl)silyl]oxy]phenyl]ethanol
Molecular Weight (g/mol) 504.867
Formula Weight 504.328
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.