Learn More
N-Despropyl-macitentan, TRC
Product Code. 30134475
Change view
Click to view available options
Quantity:
1 mg
5 mg
50 mg
Unit Size:
1mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134475 5 mg 5mg
30135960 50 mg 50mg
30008493 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134475 Supplier Toronto Research Chemicals Supplier No. TRCD2973055MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1103522-45-7
Molecular Formula C16H14Br2N6O4S
Molecular Weight (g/mol) 546.19
Synonym N-[5-(4-Bromophenyl)-6-[2-[(5-bromo-2-pyrimidinyl)oxy]ethoxy]-4-pyrimidinyl]-sulfamide, ACT-132577
SMILES O=S(N)(NC1=NC=NC(OCCOC2=NC=C(Br)C=N2)=C1C3=CC=C(Br)C=C3)=O
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C16H14Br2N6O4S/c17-11-3-1-10(2-4-11)13-14(24-29(19,25)26)22-9-23-15(13)27-5-6-28-16-20-7-12(18)8-21-16/h1-4,7-9H,5-6H2,(H2,19,25,26)(H,22,23,24)
Formula Weight 543.9164
Chemical Name or Material N-Despropyl Macitentan