missing translation for 'onlineSavingsMsg'
Learn More

N-(Diphenylmethylene)glycine tert-butyl ester, 98%

Product Code. 10502433 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10502433 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10502433 Supplier Thermo Scientific Acros Supplier No. 369320010
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 81477-94-3 | C19H21NO2 | 295.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 81477-94-3
Melting Point 111.0°C to 115.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C19H21NO2
Linear Formula (C6H5)2C=NCH2CO2C(CH3)3
MDL Number MFCD00134280
Quantity 1 g
Synonym n-diphenylmethylene glycine tert-butyl ester, diphenylmethylene-glycine t-butyl ester, tert-butyl 2-diphenylmethylene amino acetate, n-diphenylmethylene glycerine tert-butyl ester, tert-butyl 2-diphenylmethyleneamino acetate, diphenylmethyleneamino acetic acid tert-butyl ester, tert-butyl diphenylmethyleneamino acetate, n-diphenylmethylene glycinetert-butylester, tert-butyl 2-benzhydrylideneamino acetate, glycine, n-diphenylmethylene-, 1,1-dimethylethyl ester
InChI Key YSHDPXQDVKNPKA-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2
IUPAC Name tert-butyl 2-(benzhydrylideneamino)acetate
Molecular Weight (g/mol) 295.38
PubChem CID 688171
Formula Weight 295.38
Percent Purity 98%
Chemical Name or Material N-(Diphenylmethylene)glycine tert-butyl ester
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.