missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-N-nitroso-2-propanamine (100μg/mL in Methanol), TRC

Product Code. 18001510
Change view
Click to view available options
Quantity:
1 x 1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18001510 1 x 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18001510 Supplier Toronto Research Chemicals Supplier No. TRCKIT83651X1ML
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 16339-04-1
Molecular Formula C5 H12 N2 O
Quantity 1 x 1 mL
Synonym 2-Propanamine, N-ethyl-N-nitroso-, Diethylamine, 1-methyl-N-nitroso- (7CI,8CI), N-Ethyl-N-nitroso-2-propanamine, 1-Methyl-N-nitrosodiethylamine, Ethylisopropylnitrosamine, EIPNA, N-Nitroso-N-ethyl-N-isopropylamine, NEIPA, NEPA
InChI Formula InChI=1S/C5H12N2O/c1-4-7(6-8)5(2)3/h5H,4H2,1-3H3
SMILES CCN(N=O)C(C)C
IUPAC Name N-ethyl-N-propan-2-ylnitrous amide
Molecular Weight (g/mol) 116.1616
Formula Weight 116.095
Concentration or Composition (by Analyte or Components) N-Ethyl-N-nitroso-2-propanamine 100 μg/mL
Chemical Name or Material N-Ethyl-N-nitroso-2-propanamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.