missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-4-methoxy-L-phenylalanine, 95%

Product Code. 15488637
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488637 1 g 1g
15408647 250 mg 250mg
15498637 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15488637 Supplier Thermo Scientific Alfa Aesar Supplier No. H52117.03
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 77128-72-4 | C25H23NO5 | 417.461 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77128-72-4
Molecular Formula C25H23NO5
Molecular Weight (g/mol) 417.461
MDL Number MFCD00153368
InChI Key JYQODLWFOPCSCS-QHCPKHFHSA-N
Synonym fmoc-tyr me-oh, fmoc-o-methyl-l-tyrosine, fmoc-4-methoxy-l-phenylalanine, fmoc-l-4-methoxyphenylalanine, fmoc-4-methoxy-phe-oh, fmoc-o-methyl-tyr-oh, fmoc-l-tyr me, fmoc-4-methoxy-l-phe-oh, fmoc-l-4-methoxyphe
PubChem CID 2762281
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-methoxyphenyl)propanoic acid
SMILES COC1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Quantity 1 g
Formula Weight 417.45
Percent Purity 95%
Chemical Name or Material N-Fmoc-4-methoxy-L-phenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.