missing translation for 'onlineSavingsMsg'
Learn More

N'-Formylkynurenine, TRC

Product Code. 30135478
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135478 25 mg 25mg
30010158 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135478 Supplier Toronto Research Chemicals Supplier No. TRCF70049025MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1022-31-7
Molecular Formula C11 H12 N2 O4
Molecular Weight (g/mol) 236.22
Synonym α-Amino-2-(formylamino)-γ-oxo-Benzenebutanoic Acid;
IUPAC Name 2-amino-4-(2-formamidophenyl)-4-oxobutanoic acid
SMILES NC(CC(=O)c1ccccc1NC=O)C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C11H12N2O4/c12-8(11(16)17)5-10(15)7-3-1-2-4-9(7)13-6-14/h1-4,6,8H,5,12H2,(H,13,14)(H,16,17)
Formula Weight 236.0797
Percent Purity >95%
Chemical Name or Material N-Formylkynurenine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.